siRNA Details
| Gene Symbol: Gm13974 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK043709.1 | - | 9 | 1424 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB265936.2 | - | 4 | 636 | Chr. 2 - 114975470 - 115043961 [+] on Build GRCm38 | |

Gm13974
Genomic Map