siRNA Details
| Gene Symbol: Gm4117 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK087351.1 | - | 698 | |||
| BB536129.2 | - | 630 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm4117| Gene Symbol: Gm4117 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK087351.1 | - | 698 | |||
| BB536129.2 | - | 630 | ||||