siRNA Details
| Gene Symbol: Gm4117 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK087351.1 | - | 263 | |||
| BB536129.2 | - | 195 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB664604.1 | - | 2 | 263 | Chr. 13 - 89739464 - 89887078 [+] on Build GRCm38 | |

Gm4117
Genomic Map