siRNA Details
Gene Symbol: Zfp141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC063263.1 | - | 5 | 616 | ||
BC089361.1 | - | 3 | 315 | |||
CN459213.1 | - | 3 | 186 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CF735086.1 | - | 3 | 317 | Chr. 7 - 42476489 - 42490160 [-] on Build GRCm38 |
