siRNA Details
Gene Symbol: Zfp141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC063263.1 | - | 7 | 1049 | ||
BC089361.1 | - | 5 | 748 | |||
CF727770.1 | - | 1 | 199 | |||
CN459213.1 | - | 5 | 619 |
Gene Symbol: 2610021A01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK011498.1 | - | 6 | 1003 |
