siRNA Details
| Gene Symbol: Zfp141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC063263.1 | - | 7 | 1216 | ||
| BC089361.1 | - | 5 | 915 | |||
| CF727770.1 | - | 1 | 366 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp141| Gene Symbol: Zfp141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC063263.1 | - | 7 | 1216 | ||
| BC089361.1 | - | 5 | 915 | |||
| CF727770.1 | - | 1 | 366 | |||