siRNA Details
| Gene Symbol: Zfp141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC063263.1 | - | 7 | 1276 | ||
| BC089361.1 | - | 5 | 952 | |||
| CF727770.1 | - | 1 | 403 | |||
| Gene Symbol: 2610021A01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011498.1 | - | 6 | 1207 | ||

Zfp141
Genomic Map