siRNA Details
| Gene Symbol: 1700084F23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045965.1 | 5 | 794 | |||
| GenBank mRNA | AK006991.1 | - | 5 | 795 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707049.1 | - | 5 | 791 | Chr. 13 - 70004206 - 70028345 [-] on Build GRCm38 | |

1700084F23Rik
Genomic Map