siRNA Details
| Gene Symbol: 1700084F23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045965.1 | 5 | 815 | |||
| GenBank mRNA | AK006991.1 | - | 5 | 816 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700084F23Rik| Gene Symbol: 1700084F23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045965.1 | 5 | 815 | |||
| GenBank mRNA | AK006991.1 | - | 5 | 816 | ||