siRNA Details
Gene Symbol: 1700065I16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040315.2 | 3 | 348 | |||
NR_108057.1 | 4 | 378 | ||||
GenBank mRNA | AK006996.1 | - | 4 | 362 | ||
AK018879.1 | - | 3 | 350 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700065I16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040315.2 | 3 | 348 | |||
NR_108057.1 | 4 | 378 | ||||
GenBank mRNA | AK006996.1 | - | 4 | 362 | ||
AK018879.1 | - | 3 | 350 |