siRNA Details
| Gene Symbol: 1700065I16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040315.2 | 3 | 535 | |||
| NR_108057.1 | 4 | 565 | ||||
| GenBank mRNA | AK006996.1 | - | 4 | 549 | ||
| AK018879.1 | - | 3 | 537 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700065I16Rik| Gene Symbol: 1700065I16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040315.2 | 3 | 535 | |||
| NR_108057.1 | 4 | 565 | ||||
| GenBank mRNA | AK006996.1 | - | 4 | 549 | ||
| AK018879.1 | - | 3 | 537 | |||