siRNA Details
| Gene Symbol: A230059G12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK038745.1 | - | 1 | 361 | ||
| AK054148.1 | - | 1 | 1482 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A230059G12Rik| Gene Symbol: A230059G12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK038745.1 | - | 1 | 361 | ||
| AK054148.1 | - | 1 | 1482 | |||