siRNA Details
| Gene Symbol: 2610037D02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040423.1 | 1 | 21 | |||
| GenBank mRNA | AK049239.1 | - | 1 | 22 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB651835.1 | - | 1 | 22 | Chr. 15 - 96193832 - 96197974 [-] on Build GRCm38 | |

2610037D02Rik
Genomic Map