siRNA Details
| Gene Symbol: 2610037D02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040423.1 | 6 | 3072 | |||
| GenBank mRNA | AK049239.1 | - | 6 | 3073 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2610037D02Rik| Gene Symbol: 2610037D02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040423.1 | 6 | 3072 | |||
| GenBank mRNA | AK049239.1 | - | 6 | 3073 | ||