siRNA Details
Gene Symbol: 9430041J12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033568.1 | 8 | 1546 | |||
GenBank mRNA | AK051461.1 | - | 7 | 1134 | ||
AK082200.1 | - | 8 | 1548 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 9430041J12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033568.1 | 8 | 1546 | |||
GenBank mRNA | AK051461.1 | - | 7 | 1134 | ||
AK082200.1 | - | 8 | 1548 |