siRNA Details
| Gene Symbol: 9430041J12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033568.1 | 4 | 1062 | |||
| GenBank mRNA | AK020464.1 | - | 3 | 648 | ||
| AK051461.1 | - | 3 | 650 | |||
| AK082200.1 | - | 4 | 1064 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9430041J12Rik| Gene Symbol: 9430041J12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033568.1 | 4 | 1062 | |||
| GenBank mRNA | AK020464.1 | - | 3 | 648 | ||
| AK051461.1 | - | 3 | 650 | |||
| AK082200.1 | - | 4 | 1064 | |||