siRNA Details
| Gene Symbol: 4930453L07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_073360.1 | 2 | 1016 | |||
| GenBank mRNA | AK015455.1 | - | 2 | 1016 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930453L07Rik| Gene Symbol: 4930453L07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_073360.1 | 2 | 1016 | |||
| GenBank mRNA | AK015455.1 | - | 2 | 1016 | ||