siRNA Details
| Gene Symbol: 4930453O03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015456.1 | - | 260 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715004.1 | - | 3 | 260 | Chr. 1 - 88347799 - 88352930 [-] on Build GRCm38 | |

4930453O03Rik
Genomic Map