siRNA Details
| Gene Symbol: 4930453H23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015451.1 | - | 5 | 1384 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930453H23Rik| Gene Symbol: 4930453H23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015451.1 | - | 5 | 1384 | ||