siRNA Details
| Gene Symbol: 4930453J04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015453.1 | - | 1 | 1951 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY712386.1 | - | 1 | 457 | Chr. 17 - 86231979 - 86232657 [+] on Build GRCm38 | |

4930453J04Rik
Genomic Map