siRNA Details
| Gene Symbol: 4930455B14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045968.1 | 4 | 364 | |||
| GenBank mRNA | AK015458.1 | - | 4 | 309 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715008.1 | - | 309 | Chr. - - [-] on Build | ||

4930455B14Rik
Genomic Map