siRNA Details
Gene Symbol: Tmem207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK142524.1 | - | 3 | 448 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB660902.1 | - | 3 | 448 | Chr. 16 - 26517430 - 26526769 [-] on Build GRCm38 |
