siRNA Details
| Gene Symbol: Gdpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076774.1 | - | 1 | 14 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB615240.1 | - | 1 | 14 | Chr. 11 - 87074257 - 87074958 [-] on Build GRCm38 | |

Gdpd1
Genomic Map