siRNA Details
| Gene Symbol: Ipmk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076771.1 | - | 490 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV264160.2 | - | 3 | 515 | Chr. 10 - 71381749 - 71433180 [+] on Build GRCm38 | |

Ipmk
Genomic Map