siRNA Details
| Gene Symbol: 9430085M18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020506.1 | - | 1 | 374 | ||
| AK162370.1 | - | 1 | 393 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY721108.1 | - | 1 | 389 | Chr. 5 - 101703589 - 101705350 [+] on Build GRCm38 | |

9430085M18Rik
Genomic Map