siRNA Details
| Gene Symbol: 2610001N11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011267.1 | - | 1 | 596 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CB850204.2 | - | 272 | Chr. - - [-] on Build | ||

2610001N11Rik
Genomic Map