siRNA Details
| Gene Symbol: 4933425B07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046460.1 | 5 | 1686 | |||
| GenBank mRNA | AK016907.1 | - | 5 | 1687 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933425B07Rik| Gene Symbol: 4933425B07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046460.1 | 5 | 1686 | |||
| GenBank mRNA | AK016907.1 | - | 5 | 1687 | ||