siRNA Details
Gene Symbol: Kmt2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020646.1 | - | 1 | 764 | ||
AK082403.1 | - | 1 | 1072 | |||
AK154113.1 | - | 1 | 510 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Kmt2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020646.1 | - | 1 | 764 | ||
AK082403.1 | - | 1 | 1072 | |||
AK154113.1 | - | 1 | 510 |