siRNA Details
| Gene Symbol: Gm13630 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK034270.1 | - | 1, 2 | 38 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB626272.1 | - | 1, 2 | 38 | Chr. 2 - 66409495 - 66413899 [+] on Build GRCm38 | |

Gm13630
Genomic Map