siRNA Details
| Gene Symbol: Sh3glb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK052668.1 | - | 1 | 530 | ||
| AK086477.1 | - | 1 | 470 | |||
| AK147816.1 | - | 1 | 507 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sh3glb1| Gene Symbol: Sh3glb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK052668.1 | - | 1 | 530 | ||
| AK086477.1 | - | 1 | 470 | |||
| AK147816.1 | - | 1 | 507 | |||