siRNA Details
| Gene Symbol: 4933411E08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045342.1 | 4 | 1313 | |||
| GenBank mRNA | AK016769.1 | - | 4 | 1314 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933411E08Rik| Gene Symbol: 4933411E08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045342.1 | 4 | 1313 | |||
| GenBank mRNA | AK016769.1 | - | 4 | 1314 | ||