siRNA Details
| Gene Symbol: 1700101I19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007108.1 | - | 1 | 406 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707181.1 | - | 1 | 399 | Chr. 1 - 34579202 - 34579610 [-] on Build GRCm38 | |

1700101I19Rik
Genomic Map