siRNA Details
Gene Symbol: Gm4211 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK033894.1 | - | 253 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB625712.1 | - | 1, 2 | 253 | Chr. 13 - 80959210 - 80963730 [-] on Build GRCm38 |
