siRNA Details
| Gene Symbol: Gm4211 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK033894.1 | - | 94 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB625712.1 | - | 1 | 94 | Chr. 13 - 80959210 - 80963730 [-] on Build GRCm38 | |

Gm4211
Genomic Map