siRNA Details
| Gene Symbol: Gm33255 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK135250.1 | - | 3 | 599 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CJ051271.1 | - | 3 | 599 | Chr. 16 - 88368238 - 88381696 [-] on Build GRCm38 | |

Gm33255
Genomic Map