siRNA Details
| Gene Symbol: 4930577H14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019813.1 | - | 558 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715925.1 | - | 2 | 558 | Chr. 4 - 91939001 - 91939952 [-] on Build GRCm38 | |

4930577H14Rik
Genomic Map