siRNA Details
| Gene Symbol: Mcu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK166044.1 | - | 4 | 666 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mcu| Gene Symbol: Mcu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK166044.1 | - | 4 | 666 | ||