siRNA Details
| Gene Symbol: Pdpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK086050.1 | - | 1 | 412 | ||
| AK160524.1 | - | 1 | 456 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG916039.1 | - | 1 | 58 | Chr. 8 - 111137265 - 111137978 [+] on Build GRCm38 | |

Pdpr
Genomic Map