siRNA Details
| Gene Symbol: Pdpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK079427.1 | - | 1 | 645 | ||
| AK086050.1 | - | 1 | 1714 | |||
| AK160524.1 | - | 1 | 1749 | |||
| BC064088.1 | - | 1 | 428 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdpr| Gene Symbol: Pdpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK079427.1 | - | 1 | 645 | ||
| AK086050.1 | - | 1 | 1714 | |||
| AK160524.1 | - | 1 | 1749 | |||
| BC064088.1 | - | 1 | 428 | |||