siRNA Details
| Gene Symbol: 5430440L12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017417.1 | - | 1 | 1001 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV478375.1 | - | 1 | 207 | Chr. 19 - 4794739 - 4795162 [-] on Build GRCm38 | |

5430440L12Rik
Genomic Map