siRNA Details
| Gene Symbol: A230106O10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020718.1 | - | 1 | 491 | ||
| BB284628.1 | - | 1 | 370 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A230106O10Rik| Gene Symbol: A230106O10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020718.1 | - | 1 | 491 | ||
| BB284628.1 | - | 1 | 370 | |||