siRNA Details
| Gene Symbol: 2810029C07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045295.1 | 1 | 416 | |||
| GenBank mRNA | AK012841.1 | - | 1 | 353 | ||
| AK048965.1 | - | 371 | ||||
| BC024729.1 | - | 1 | 416 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2810029C07Rik| Gene Symbol: 2810029C07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045295.1 | 1 | 416 | |||
| GenBank mRNA | AK012841.1 | - | 1 | 353 | ||
| AK048965.1 | - | 371 | ||||
| BC024729.1 | - | 1 | 416 | |||