siRNA Details
| Gene Symbol: 4930579K21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076967.1 | - | 2, 3 | 420 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB615956.1 | - | 2, 3 | 418 | Chr. 11 - 50723774 - 50725542 [-] on Build GRCm38 | |

4930579K21
Genomic Map