siRNA Details
| Gene Symbol: 4930579K21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076967.1 | - | 6 | 1247 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930579K21| Gene Symbol: 4930579K21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076967.1 | - | 6 | 1247 | ||