siRNA Details
| Gene Symbol: LOC102640562 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK084494.1 | - | 6 | 778 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB474543.2 | - | 3 | 416 | Chr. 2 - 106642921 - 106646479 [-] on Build GRCm38 | |

LOC102640562
Genomic Map