siRNA Details
| Gene Symbol: A930016I07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020866.1 | - | 1 | 626 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB283723.1 | - | 1 | 281 | Chr. 7 - 114364336 - 114364872 [-] on Build GRCm38 | |

A930016I07Rik
Genomic Map