siRNA Details
| Gene Symbol: 4921532D01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014993.1 | - | 1 | 1116 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4921532D01Rik| Gene Symbol: 4921532D01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014993.1 | - | 1 | 1116 | ||