siRNA Details
| Gene Symbol: Gm12126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK139851.1 | - | 2 | 604 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY725970.1 | - | 2 | 603 | Chr. 11 - 36080664 - 36083910 [+] on Build GRCm38 | |

Gm12126
Genomic Map