siRNA Details
| Gene Symbol: 4930401A07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015026.1 | - | 4 | 648 | ||
| AV258591.1 | - | 1 | 196 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930401A07Rik| Gene Symbol: 4930401A07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015026.1 | - | 4 | 648 | ||
| AV258591.1 | - | 1 | 196 | |||