siRNA Details
| Gene Symbol: 4922502B01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015021.1 | - | 1033 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB679327.1 | - | 1 | 229 | Chr. 8 - 109250481 - 109250925 [-] on Build GRCm38 | |

4922502B01Rik
Genomic Map